Assay Details
| Gene Symbol: Prrt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_077057.1 | 1 - 2 | 110 | 87 | ||
| GenBank mRNA | CB738953.1 | - | 1 - 2 | 109 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Paxip1-associated glutamate-rich protein 1 | Pagr1 |
Pa1, RGD1305592 |
-1.6kb | Sense | Chr. 1 - 198448790 - 198451078 on NCBI Build Rnor_6.0 |
| Downstream | MYC-associated zinc finger protein (purine-binding transcription factor) | Maz | Pur-1 | 2.0kb | Sense | Chr. 1 - 198456699 - 198460126 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

Prrt2
Genomic Map