Assay Details
| Gene Symbol: Mir3075 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106690.1 | 1 | 61 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102553140 | LOC102553140 | -70.3kb | Sense | Chr. 16 - 1749528 - 1752476 on NCBI Build Rnor_6.0 | |
| Downstream | peptidylprolyl isomerase F | Ppif |
CyP-D, CypD, PPIase |
156.3kb | Sense | Chr. 16 - 1979191 - 1985912 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3075
Genomic Map