Assay Details
| Gene Symbol: Mir6329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106701.1 | 1 | 46 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | radixin | Rdx | -16.7kb | Anti-sense | Chr. 8 - 56570728 - 56612851 on NCBI Build Rnor_6.0 | |
| Downstream | zinc finger CCCH type containing 12C | Zc3h12c | RGD1565078 | 9.4kb | Sense | Chr. 8 - 56639047 - 56696968 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6329
Genomic Map