Assay Details
Gene Symbol: LOC100910558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_110678.1 | 1 - 2 | 272 | 87 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DY316388.1 | - | 164 | Chr. - - [-] on Build | 87 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear factor I/B | Nfib | -2.5kb | Anti-sense | Chr. 5 - 100433268 - 100647963 on NCBI Build Rnor_6.0 | |
Downstream |
zinc finger, DHHC-type containing 21 |
Zdhhc21 | 241.6kb | Anti-sense | Chr. 5 - 100924621 - 100983380 on NCBI Build Rnor_6.0 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
