Assay Details
| Gene Symbol: LOC100910558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110680.1 | 1 - 2 | 278 | 66 | ||
| GenBank mRNA | FM089179.1 | - | 1 - 2 | 271 | 66 | |
| FM099320.1 | - | 1 - 2 | 229 | 66 | ||
| FQ215485.1 | - | 1 - 2 | 278 | 66 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear factor I/B | Nfib | -2.5kb | Anti-sense | Chr. 5 - 100433268 - 100647963 on NCBI Build Rnor_6.0 | |
| Downstream |
zinc finger, DHHC-type containing 21 |
Zdhhc21 | 236.4kb | Anti-sense | Chr. 5 - 100924621 - 100983380 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

LOC100910558
Genomic Map