Assay Details
| Gene Symbol: LOC100910237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110684.1 | 1 - 2 | 365 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
solute carrier family 18, subfamily B, member 1 |
Slc18b1 | RGD1306962 | -8.7kb | Sense | Chr. 1 - 22705980 - 22748559 on NCBI Build Rnor_6.0 |
| Downstream | ribosomal protein S12 | Rps12 | 186.0 | Anti-sense | Chr. 1 - 22758214 - 22760370 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

LOC100910237
Genomic Map