Assay Details
| Gene Symbol: LOC100909970 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110686.1 | 2 - 3 | 415 | 68 | ||
| GenBank mRNA | FQ230299.1 | - | 2 - 3 | 415 | 68 | |
| FQ232833.1 | - | 2 - 3 | 413 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ras responsive element binding protein 1 | Rreb1 | -122.4kb | Anti-sense | Chr. 17 - 27539137 - 27718898 on NCBI Build Rnor_6.0 | |
| Downstream | neuritin 1 | Nrn1 | Nrn | 75.1kb | Anti-sense | Chr. 17 - 27925054 - 28002946 on NCBI Build Rnor_6.0 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100909970
Genomic Map