Assay Details
| Gene Symbol: Mir3102 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106684.1 | 1 | 92 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RELT tumor necrosis factor receptor | Relt | Tnfrsf19l | -19.2kb | Sense | Chr. 1 - 165884407 - 165902012 on NCBI Build Rnor_6.0 |
| Downstream |
pyrimidinergic receptor P2Y, G-protein coupled, 6 |
P2ry6 | P2y6 | 51.1kb | Sense | Chr. 1 - 165972439 - 166008351 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir3102
Genomic Map