Assay Details
| Gene Symbol: Mir6321 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106692.1 | 1 | 93 | 134 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RUN and FYVE domain containing 3 | Rufy3 |
Ripx, Singar1 |
Sense | Chr. 14 - 21058575 - 21144743 on NCBI Build Rnor_6.0 | |
| Downstream |
UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae) |
Utp3 | Crlz1 | 56.5kb | Sense | Chr. 14 - 21153998 - 21155652 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6321
Genomic Map