Assay Details
| Gene Symbol: Mir6324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106696.1 | 1 | 66 | 89 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | bromodomain and WD repeat domain containing 1 | Brwd1 | Wdr9 | Sense | Chr. 11 - 36338080 - 36430474 on NCBI Build Rnor_6.0 | |
| Downstream | high mobility group nucleosome binding domain 1 | Hmgn1 | 90.4kb | Sense | Chr. 11 - 36474028 - 36479947 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mir6324
Genomic Map