Assay Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277228.1 | NP_001264157.1 | 8 - 9 | 1651 | 96 | |
| GenBank mRNA | BC085943.1 | - | 7 - 8 | 1771 | 96 | |
| BC168987.1 | - | 8 - 9 | 1613 | 9030170 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdelc1
Genomic Map