Assay Details
| Gene Symbol: Nlrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228200.2 | XP_006228262.1 | 5 - 6 | 2194 | 61 | |
| XM_006228201.2 | XP_006228263.1 | 5 - 6 | 2237 | 61 | ||
More Information
Set Membership: |
|

Nlrp4
Genomic Map