Assay Details
| Gene Symbol: Tubg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145778.2 | NP_665721.1 | 7 - 8 | 810 | 58 | |
| GenBank mRNA | AB015946.1 | - | 7 - 8 | 715 | 58 | |
| BC070957.1 | - | 7 - 8 | 810 | 7105643 | 58 | |
| Gene Symbol: Tubg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191075.1 | NP_001178004.1 | 8 - 9 | 757 | 58 | |
| XM_006247427.2 | XP_006247489.1 | 7 - 8 | 780 | 58 | ||
More Information
Set Membership: |
|

Tubg1
Genomic Map