Assay Details
| Gene Symbol: Mir421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_031947.1 | 1 | 39 | 73 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102554630 | LOC102554630 | -96.4kb | Anti-sense | Chr. X - 74337707 - 74347101 on NCBI Build Rnor_6.0 | |
| Downstream | microRNA mir-374 | Mir374 | rno-mir-374 | 78.0 | Sense | Chr. X - 74443648 - 74443715 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

Mir421
Genomic Map