Assay Details
Gene Symbol: Mir1249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_037390.1 | 1 | 55 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear pore associated protein | Npap60 |
Nup50, Rtp60 |
-10.4kb | Anti-sense | Chr. 7 - 125833557 - 125850830 on NCBI Build Rnor_6.0 |
Downstream | uroplakin 3A | Upk3a | 59.0kb | Anti-sense | Chr. 7 - 125920357 - 125925430 on NCBI Build Rnor_6.0 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
