Assay Details
| Gene Symbol: RGD1564124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FM112750.1 | - | 4 - 5 | 345 | 82 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-374 | Mir374 | rno-mir-374 | -31.0kb | Anti-sense | Chr. X - 74443648 - 74443715 on NCBI Build Rnor_6.0 |
| Downstream |
zinc finger, CCHC domain containing 13 |
Zcchc13 | 22.1kb | Sense | Chr. X - 74497112 - 74498299 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

RGD1564124
Genomic Map