Assay Details
| Gene Symbol: LOC100912266 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY387089.1 | - | 1 - 2 | 80 | 105 | |
| AY539874.1 | - | 1 - 2 | 80 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | phosphodiesterase 8B | Pde8b | RNPDE8B | -83.5kb | Anti-sense | Chr. 2 - 24718548 - 24923718 on NCBI Build Rnor_6.0 |
| Downstream |
zinc finger, BED-type containing 3 |
Zbed3 | 28.1kb | Sense | Chr. 2 - 25039497 - 25052129 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

LOC100912266
Genomic Map