Assay Details
| Gene Symbol: Cysltr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008770842.1 | XP_008769064.1 | 1 - 2 | 361 | 102 | |
| XM_008770843.1 | XP_008769065.1 | 2 - 3 | 1791 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | fibronectin type III domain containing 3a | Fndc3a | Fndc3 | -325.3kb | Sense | Chr. 15 - 54378865 - 54582959 on NCBI Build Rnor_6.0 |
| Downstream | similar to mitochondrial ribosomal protein L40 | LOC684557 | 43.7kb | Anti-sense | Chr. 15 - 54954017 - 54961713 on NCBI Build Rnor_6.0 |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008770842.1, XM_008770843.1 transcripts.
Set Membership: |
|

Cysltr2
Genomic Map