Assay Details
| Gene Symbol: Mir6330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_106702.1 | 1 | 79 | 119 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | similar to Nucleoside diphosphate-linked moiety X motif 16 (Nudix motif 16) | LOC688828 | -28.5kb | Sense | Chr. 8 - 113687983 - 113689657 on NCBI Build Rnor_6.0 | |
| Downstream | asteroid homolog 1 (Drosophila) | Aste1 | RGD1304643 | 218.7kb | Anti-sense | Chr. 8 - 113936991 - 113954969 on NCBI Build Rnor_6.0 |
More Information
Set Membership: |
|

Mir6330
Genomic Map