Assay Details
| Gene Symbol: Lrrgt00014 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U83119.1 | - | 1 | 2497 | 109 | |
| Gene Symbol: LOC499182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100474.1 | - | 3 | 4193 | 109 | |
| Gene Symbol: LOC499355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100475.1 | - | 3 | 4190 | 109 | |
| Gene Symbol: LOC501503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100477.1 | - | 2 | 4194 | 109 | |
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY539942.1 | - | 109 | |||
More Information
Set Membership: |
|

Lrrgt00014
Genomic Map