Assay Details
| Gene Symbol: Mbnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106949.1 | NP_001100419.1 | 2 | 338 | 86 | |
| XM_008773556.1 | XP_008771778.1 | 2 | 1817 | 86 | ||
| XM_008773557.1 | XP_008771779.1 | 2 | 940 | 86 | ||
| XM_008773558.1 | XP_008771780.1 | 2 | 937 | 86 | ||
| XM_008773559.1 | XP_008771781.1 | 2 | 940 | 86 | ||
| XM_008773560.1 | XP_008771782.1 | 2 | 943 | 86 | ||
| XM_008773561.1 | XP_008771783.1 | 2 | 940 | 86 | ||
| XM_008773562.1 | XP_008771784.1 | 2 | 213 | 86 | ||
More Information
Set Membership: |
|

Mbnl3
Genomic Map