Assay Details
| Gene Symbol: Plac8l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728543.4 | XP_002728589.1 | 1 | 468 | 99 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002728543.4 transcripts.
Set Membership: |
|

Plac8l1
Genomic Map