Assay Details
| Gene Symbol: Zfp532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006254865.2 | XP_006254927.1 | 1 | 1845 | 60 | |
| XM_006254873.2 | XP_006254935.1 | 1 | 1845 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006254865.2, XM_006254873.2 transcripts.
Set Membership: |
|

Zfp532
Genomic Map