Assay Details
Gene Symbol: Dynap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006254931.2 | XP_006254993.1 | 3 - 4 | 572 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
