Assay Details
| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047090.1 | NP_001040555.1 | 6 | 1254 | 99 | |
| GenBank mRNA | DQ672552.1 | - | 6 | 1254 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047090.1 | NP_001040555.1 | 6 | 1254 | 99 | |
| GenBank mRNA | DQ672552.1 | - | 6 | 1254 | 99 | |
Set Membership: |
|