Assay Details
| Gene Symbol: Chst4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006255586.2 | XP_006255648.1 | 2 | 2661 | 88 | |
| XM_006255587.2 | XP_006255649.1 | 2 | 2744 | 88 | ||
More Information
Set Membership: |
|

Chst4
Genomic Map