Assay Details
| Gene Symbol: LOC100366044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760951.1 | XP_008759173.1 | 4 | 4156 | 117 | |
| XM_008760952.1 | XP_008759174.1 | 4 | 4180 | 117 | ||
More Information
Set Membership: |
|

LOC100366044
Genomic Map