Assay Details
| Gene Symbol: LOC102555481 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761449.1 | XP_008759671.1 | 2 - 3 | 210 | 63 | |
| Gene Symbol: Chi3l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191712.1 | NP_001178641.1 | 8 - 9 | 921 | 63 | |
| XM_008761383.1 | XP_008759605.1 | 8 - 9 | 921 | 63 | ||
| GenBank mRNA | JF781276.1 | - | 8 - 9 | 939 | 63 | |
More Information
Set Membership: |
|

LOC102555481
Genomic Map