Assay Details
| Gene Symbol: LOC102547192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231983.2 | XP_006232045.1 | 1 | 1710 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102547192| Gene Symbol: LOC102547192 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006231983.2 | XP_006232045.1 | 1 | 1710 | 60 | |
Set Membership: |
|