Assay Details
| Gene Symbol: Rhno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027366.1 | 3 | 1204 | 96 | ||
| NR_027367.1 | 3 | 818 | 96 | |||
| GenBank mRNA | BC079219.1 | - | 3 | 1204 | 96 | |
| Gene Symbol: LOC100362315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008761349.1 | XP_008759571.1 | 1 | 827 | 96 | |
More Information
Set Membership: |
|

Rhno1
Genomic Map