Assay Details
| Gene Symbol: Gbp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749410.3 | XP_003749458.2 | 5 - 6 | 764 | 101 | |
| XM_006233426.2 | XP_006233488.1 | 5 - 6 | 764 | 101 | ||
More Information
Set Membership: |
|

Gbp3
Genomic Map