Assay Details
| Gene Symbol: Ak6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012463.3 | NP_001012481.1 | 4 - 5 | 376 | 126 | |
| GenBank mRNA | BC089989.1 | - | 4 - 5 | 342 | 7319327 | 126 |
| BQ205289.1 | - | 1 - 2 | 126 | |||
| DQ323055.1 | - | 4 - 5 | 325 | 126 | ||
More Information
Set Membership: |
|

Ak6
Genomic Map