Assay Details
| Gene Symbol: LOC499182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100474.1 | - | 3 | 5510 | 140 | |
| Gene Symbol: LOC317588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY321321.1 | - | 2 | 938 | 140 | |
| Gene Symbol: LOC361942 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY539867.1 | - | 1462 | 140 | ||
| Gene Symbol: LOC499355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100475.1 | - | 3 | 5507 | 140 | |
| Gene Symbol: LOC501503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ100477.1 | - | 2 | 5507 | 140 | |
| Gene Symbol: Serinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY539942.1 | - | 3109 | 140 | ||
| Gene Symbol: Vps13a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY325251.1 | - | 10 | 2495 | 140 | |
More Information
Set Membership: |
|

LOC499182
Genomic Map