Assay Details
| Gene Symbol: LOC102554393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233879.2 | XP_006233941.2 | 16 - 17 | 2049 | 101 | |
| Gene Symbol: LOC102554283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233729.2 | XP_006233791.1 | 1 - 2 | 817 | 101 | |
More Information
Set Membership: |
|

LOC102554393
Genomic Map