Assay Details
| Gene Symbol: LOC102554393 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233879.2 | XP_006233941.2 | 18 - 19 | 2322 | 90 | |
| Gene Symbol: LOC102554283 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006233729.2 | XP_006233791.1 | 3 - 4 | 1090 | 90 | |
More Information
Set Membership: |
|

LOC102554393
Genomic Map