Assay Details
| Gene Symbol: Dscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108316.2 | NP_001101786.2 | 8 | 1907 | 103 | |
| XM_006248072.2 | XP_006248134.1 | 7 | 2211 | 103 | ||
| XM_006248073.2 | XP_006248135.1 | 7 | 2209 | 103 | ||
| XM_008768583.1 | XP_008766805.1 | 8 | 2295 | 103 | ||
| GenBank mRNA | BC160826.1 | - | 8 | 1907 | 7444837 | 103 |
More Information
Set Membership: |
|

Dscr3
Genomic Map