Assay Details
| Gene Symbol: Kcng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106545.1 | NP_001100015.1 | 2 | 557 | 82 | |
| XM_008762484.1 | XP_008760706.1 | 2 | 376 | 82 | ||
| GenBank mRNA | M81784.1 | - | 2 | 382 | 82 | |
More Information
Set Membership: |
|

Kcng1
Genomic Map