Assay Details
| Gene Symbol: LOC103693464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768305.1 | XP_008766527.1 | 2 - 3 | 149 | 116 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768305.1 transcripts.
Set Membership: |
|

LOC103693464
Genomic Map