Assay Details
| Gene Symbol: LOC103693464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768305.1 | XP_008766527.1 | 23 - 24 | 1839 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103693464| Gene Symbol: LOC103693464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008768305.1 | XP_008766527.1 | 23 - 24 | 1839 | 79 | |
Set Membership: |
|