Assay Details
| Gene Symbol: LOC100911434 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762451.1 | XP_008760673.1 | 13 - 14 | 1456 | 73 | |
| Gene Symbol: Fer1l4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106534.2 | NP_001100004.2 | 14 - 15 | 1489 | 73 | |
More Information
Set Membership: |
|

LOC100911434
Genomic Map