Assay Details
| Gene Symbol: Ppp1r15b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249781.2 | XP_006249843.1 | 1 | 704 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006249781.2 transcripts.
Set Membership: |
|

Ppp1r15b
Genomic Map