Assay Details
| Gene Symbol: Rab22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108966.1 | NP_001102436.1 | 6 | 775 | 63 | |
| XM_006235705.2 | XP_006235767.1 | 7 | 951 | 63 | ||
| GenBank mRNA | BC158650.1 | - | 7 | 775 | 8367507 | 63 |
More Information
Set Membership: |
|

Rab22a
Genomic Map