Assay Details
| Gene Symbol: LOC684545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759087.1 | XP_008757309.1 | 7 - 8 | 2663 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC684545| Gene Symbol: LOC684545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759087.1 | XP_008757309.1 | 7 - 8 | 2663 | 93 | |
Set Membership: |
|