Assay Details
| Gene Symbol: Sult2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131903.1 | NP_571978.1 | 3 - 4 | 506 | 85 | |
| XM_008758891.1 | XP_008757113.1 | 3 - 4 | 536 | 85 | ||
| XM_008758892.1 | XP_008757114.1 | 3 - 4 | 535 | 85 | ||
| XM_008758893.1 | XP_008757115.1 | 3 - 4 | 524 | 85 | ||
| GenBank mRNA | D14987.1 | - | 3 - 4 | 506 | 85 | |
| D14988.1 | - | 3 - 4 | 506 | 85 | ||
| FQ209430.1 | - | 3 - 4 | 381 | 85 | ||
| FQ209460.1 | - | 3 - 4 | 381 | 85 | ||
| FQ209972.1 | - | 3 - 4 | 381 | 85 | ||
| FQ210003.1 | - | 3 - 4 | 381 | 85 | ||
| FQ210538.1 | - | 3 - 4 | 381 | 85 | ||
| FQ218232.1 | - | 3 - 4 | 381 | 85 | ||
| FQ218942.1 | - | 3 - 4 | 381 | 85 | ||
| FQ218960.1 | - | 3 - 4 | 381 | 85 | ||
| FQ218962.1 | - | 3 - 4 | 381 | 85 | ||
| FQ219069.1 | - | 3 - 4 | 381 | 85 | ||
| FQ219256.1 | - | 3 - 4 | 381 | 85 | ||
| FQ219272.1 | - | 3 - 4 | 402 | 85 | ||
| FQ219707.1 | - | 3 - 4 | 381 | 85 | ||
| M31363.1 | - | 3 - 4 | 506 | 85 | ||
More Information
Set Membership: |
|

Sult2a1
Genomic Map