Assay Details
Gene Symbol: Tnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006236605.2 | XP_006236667.1 | 5 - 6 | 408 | 113 | |
XM_006236606.2 | XP_006236668.1 | 5 - 6 | 408 | 113 | ||
XM_006236607.2 | XP_006236669.1 | 4 - 5 | 279 | 113 | ||
XM_008762999.1 | XP_008761221.1 | 4 - 5 | 360 | 113 | ||
XM_008763000.1 | XP_008761222.1 | 5 - 6 | 373 | 113 | ||
XM_008763001.1 | XP_008761223.1 | 3 - 4 | 348 | 113 | ||
XM_008763002.1 | XP_008761224.1 | 4 - 5 | 264 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
