Assay Details
| Gene Symbol: Zfp248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729406.4 | XP_002729452.2 | 4 | 2535 | 77 | |
| XM_006237213.2 | XP_006237275.1 | 5 | 2578 | 77 | ||
| XM_006237215.2 | XP_006237277.1 | 4 | 2531 | 77 | ||
| XM_008763270.1 | XP_008761492.1 | 3 | 2750 | 77 | ||
| GenBank mRNA | AY724530.1 | - | 1790 | 77 | ||
More Information
Set Membership: |
|

Zfp248
Genomic Map