Assay Details
| Gene Symbol: LOC102551565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228341.2 | XP_006228403.1 | 2 | 620 | 75 | |
| Gene Symbol: LOC102553890 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228355.2 | XP_006228417.2 | 3 | 247 | 75 | |
More Information
Set Membership: |
|

LOC102551565
Genomic Map