Assay Details
| Gene Symbol: Ifnl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228638.1 | XP_006228700.1 | 4 - 5 | 482 | 72 | |
| Gene Symbol: LOC100911249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008759287.1 | XP_008757509.1 | 4 - 5 | 482 | 72 | |
More Information
Set Membership: |
|

Ifnl3
Genomic Map