Assay Details
| Gene Symbol: Ywhaq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013053.1 | NP_037185.1 | 6 | 1112 | 76 | |
| GenBank mRNA | BC062409.1 | - | 5 | 1082 | 5623850 | 76 |
| D17614.1 | - | 6 | 1112 | 76 | ||
| FQ218679.1 | - | 5 | 872 | 76 | ||
| FQ219995.1 | - | 2 | 399 | 76 | ||
| FQ232769.1 | - | 1 | 179 | 76 | ||
More Information
Set Membership: |
|

Ywhaq
Genomic Map