Assay Details
Gene Symbol: Zfp27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008759225.1 | XP_008757447.1 | 2 - 3 | 890 | 86 | |
XM_008759226.1 | XP_008757448.1 | 2 - 3 | 278 | 86 | ||
GenBank mRNA | FQ211480.1 | - | 2 - 3 | 215 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
